1-Butanol-[D10], Sigma-Aldrich®
Catalog # SIAL302996-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:84.04 g/mol
Boiling Pt:116…118 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.92 g/cm³ (20 °C)
UN:1120
ADR:3,III
:M-Clarity™ quality level = MQ200
CAS Number:34193-38-9
Formula:DO(CD₂)₃CD₃