ContactUsLinkComponent
Multiclean® Plus Wipe Rolls
Multiclean® Plus Wipe Rolls
Catalog # ZVGA11224-00
Multiclean® Plus Wipe Rolls
Catalog # ZVGA11224-00
Specifications
  • Packed:
    500 pieces
  • Colour:
    Blue
  • Description:
    Multiclean® Plus, wipes
  • L×W:
    360×220 mm
  • Pk:
    2 Roll
Specifications
About this item
These wipes have point-to-point micro-embossing.
  • Easy to grip
  • High volume
  • Very absorbent
  • Resistant
  • High wet strength
  • Silicone-free
  • Two ply
Ideal for industry and crafts, light to coarse dirty work and metal processing and maintenance of machines.