ContactUsLinkComponent
ApoSight™ Green Caspase 3/7 substrate
ApoSight™ Green Caspase 3/7 substrate
Catalog # AATB13290
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ApoSight™ Green Caspase 3/7 substrate
Catalog # AATB13290
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    ApoSight™ Green Caspase 3/7 substrate
Specifications
About this item
ApoSight™ Green Caspase 3/7 substrate is the first fluorogenic probe for the direct fluorescence imaing of caspase activities in live cells.
  • unique fluorogenic caspase activity probe for live cells
  • sensitive and accurate detection
  • can be adapted for automation
Caution: Storage: Freeze (< −15 °C)