ContactUsLinkComponent
Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence*
Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence*
Catalog # AATB5523
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric Maleimide Quantitation Kit *Green Fluorescence*
Catalog # AATB5523
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ Fluorimetric maleimide quantitation kit *green fluorescence*
  • Tests per kit:
    200 Tests
Specifications
About this item
A variety of crosslinking reagents with a maleimide group are widely used for crosslinking proteins to proteins or proteins to other biomolecules.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.