ContactUsLinkComponent
Amplite® Fluorimetric D-Lactate Dehydrogenase (LDH) Assay Kit
Amplite® Fluorimetric D-Lactate Dehydrogenase (LDH) Assay Kit
Catalog # AATB13808
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric D-Lactate Dehydrogenase (LDH) Assay Kit
Catalog # AATB13808
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ Fluorimetric d-lactate dehydrogenase (ldh) assay kit
  • Tests per kit:
    200 Tests
Specifications
About this item
Lactate dehydrogenase (LDH) is an oxidoreductase enzyme that catalyses the interconversion of pyruvate and lactate.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.