ContactUsLinkComponent
Amplite® Luminometric Alkaline Phosphatase Assay Kit *Luminescence*
Amplite® Luminometric Alkaline Phosphatase Assay Kit *Luminescence*
Catalog # AATB11956
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Luminometric Alkaline Phosphatase Assay Kit *Luminescence*
Catalog # AATB11956
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Amplite™ Luminometric alkaline phosphatase assay kit *luminescence*
  • Tests per kit:
    100 Tests
Specifications
About this item
Alkaline phosphatase is a highly sensitive enzyme for ELISA, immuno-histochemical, Northern, Southern and Western blot applications.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.