ContactUsLinkComponent
Amplite® Fluorimetric Lysyl Oxidase Assay Kit *Red Fluorescence*
Amplite® Fluorimetric Lysyl Oxidase Assay Kit *Red Fluorescence*
Catalog # AATB15255
Supplier:  AAT Bioquest
CAS Number:  
undefined
Amplite® Fluorimetric Lysyl Oxidase Assay Kit *Red Fluorescence*
Catalog # AATB15255
Supplier:  AAT Bioquest
CAS Number:  
Specifications
  • Pk:
    500 Tests
  • Description:
    Amplite™ Fluorimetric lysyl oxidase assay kit *red fluorescence*
  • Tests per kit:
    500 Tests
Specifications
About this item
Lysyl oxidase (LOX) is an extracellular enzyme that catalyses formation of aldehydes from lysine residues in collagen and elastin precursors.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.