ContactUsLinkComponent
Cell Navigator® Mitochondrion Staining Kit *Deep Red Fluorescence*
Cell Navigator® Mitochondrion Staining Kit *Deep Red Fluorescence*
Catalog # AATB22669
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Navigator® Mitochondrion Staining Kit *Deep Red Fluorescence*
Catalog # AATB22669
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    500 Assays
  • Description:
    Cell Navigator™ Mitochondrion staining kit *Deep red fluorescence*
Specifications
About this item
The Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labeling sub-cellular organelles such as membranes, lysosomes, mitochondria and nuclei etc.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.