ContactUsLinkComponent
Cell Meter™ Fluorimetric Intracellular Peroxynitrite Assay Kit *Green Fluorescence*
Cell Meter™ Fluorimetric Intracellular Peroxynitrite Assay Kit *Green Fluorescence*
Catalog # AATB16315
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Cell Meter™ Fluorimetric Intracellular Peroxynitrite Assay Kit *Green Fluorescence*
Catalog # AATB16315
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Cell Meter™ Fluorimetric intracellular Peroxynitrite assay kit *Green fluorescence*
  • Tests per kit:
    100 Tests
Specifications
About this item
Peroxynitrite (ONOO-) is a strong oxidizing species and a highly active nitrating agent.
  • High sensitivity
  • Bright fluorescence
  • Suitable for live cell imaging
Caution: For research use only.