Order Entry
Czech Republic
ContactUsLinkComponent
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
Catalog # AATB22120
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
Catalog # AATB22120
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Description:
    C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
  • Purity:
    95%

Specifications

About this item

The green fluorescent BODIPY FL C12 fatty acid can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.

  • Phospholipid Precursor
  • pH-Insensitive Fluorescence
  • Fluorescent In Aqueous and Lipids
Caution: For research use only.