Order Entry
Czech Republic
ContactUsLinkComponent
Bodi Fluor™ R6G NHS Ester [equivalent to Bodipy® R6G, NHS Ester]
Bodi Fluor™ R6G NHS Ester [equivalent to Bodipy® R6G, NHS Ester]
Catalog # AATB703
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Bodi Fluor™ R6G NHS Ester [equivalent to Bodipy® R6G, NHS Ester]
Catalog # AATB703
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Description:
    Bodi Fluor™R6G
  • Purity:
    90%

Specifications

About this item

Bodi Fluor™R6G, NHS ester is the same molecule to Bodipy® R6G, NHS ester (Bodipy® is the trademark of Invitrogen).

  • Multiple Bioconjugation Applications
  • 90% Purity
Caution: For research use only.