Anti-IgG Goat Polyclonal Antibody [clone: IgG] (iFluor® 555)
iFluor® dyes are optimised for labeling proteins, in particular, antibodies.
- Bright signal
- Increased photostability
- Minimal quenching on proteins
: For research use only.
Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: iFluor® 555
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
- Catalog No:
- AATB16690
- AATB16831
- Symbol antigenu:
- IgG
- IgG
- Název antigenu:
- Immunoglobulin G
- Immunoglobulin G
- Druh protilátky:
- Sekundární
- Sekundární
- Klonovalita:
- Polyclonal
- Polyclonal
- Spojení:
- iFluor® 555
- iFluor® 555
- Klon:
- IgG
- IgG
- Reaktivita:
- Rabbit
- Rabbit
- Hostitel:
- Goat
- Goat
- Izotyp:
- IgG (H, L)
- IgG (H, L)
- Křížová adsorbce:
- Min×Human
- Min×Human
Frequently Bought Together