Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG Goat Polyclonal Antibody (iFluor® 633)
Anti-IgG Goat Polyclonal Antibody (iFluor® 633)
Supplier:  AAT Bioquest
Certificates
undefined
Anti-IgG Goat Polyclonal Antibody (iFluor® 633)
Supplier:  AAT Bioquest

Specifications

  • Catalog No:
  • AATB16478
  • AATB16743
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Immunoglobulin G
  • Immunoglobulin G
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • iFluor® 633
  • iFluor® 633
  • Clone:
  • IgG
  • IgG
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Isotype:
  • IgG (H, L)
  • IgG (H, L)
  • Cross adsorption:

Specifications

Frequently Bought Together