Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant PPM1B
Human Recombinant PPM1B
Catalog # ORIGTP312918
Supplier:  OriGene
CAS Number:  
Human Recombinant PPM1B
Catalog # ORIGTP312918
Supplier:  OriGene
CAS Number:  

Specifications

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    C-Myc/DDK
  • Storage conditions:
    Store at −80 °C
  • Protein synonyms:
    PP2C-beta|protein phosphatase 1B, magnesium-dependent, beta isoform|PPC2BETAX|PP2CBETA|PP2CB|PP2C-beta-X
  • UniProtKB:
    O75688
  • Protein/peptide name:
    PPM1B
  • Purity:
    >80% as determined by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    52,5 kDa
  • Sequence:
    MGAFLDKPKTEKHNAHGAGNGLRYGLSSMQGWRVEMEDAHTAVVGIPHGLEDWSFFAVYDGHAGSRVANY
    CSTHLLEHITTNEDFRAAGKSGSALELSVENVKNGIRTGFLKIDEYMRNFSDLRNGMDRSGSTAVGVMIS
    PKHIYFINCGDSRAVLYRNGQVCFSTQDHKPCNPREKERIQNAGGSVMIQRVNGSLAVSRALGDYDYKCV
    DGKGPTEQLVSPEPEVYEILRAEEDEFIILACDGIWDVMSNEELCEYVKSRLEVSDDLENVCNWVVDTCL
    HKGSRDNMSIVLVCFSNAPKVSDEAVKKDSELDKHLESRVEEIMEKSGEEGMPDLAHVMRILSAENIPNL
    PPGGGLAGKRNVIEAVYSRLNPHRESDGASDEAEESGSQGKLVEALRQMRINHRGNYRQLLEEMLTSYRL
    AKVEGEESPAEPAATATSSNSDAGNPVTMQESHTESESGLAELDSSNEDAGTKMSGEKI

    TRTRPLEQKLISEEDLAANDILDYKDDDDKV
  • Concentration:
    >0,05 µg/µl as determined by microplate BCA method

Specifications

About this item

The protein encoded by this gene is a member of the PP2C family of Ser/Thr protein phosphatases. PP2C family members are known to be negative regulators of cell stress response pathways.

  • Expression host: HEK293T
  • Buffer: 25 mM Tris-HCl, 100 mM glycine, pH 7,3, 10% glycerol

Stable for 12 months from the date of receipt of the product under proper storage and handling conditions. Avoid repeated freeze-thaw cycles.

This phosphatase has been shown to dephosphorylate cyclin-dependent kinases (CDKs), and thus may be involved in cell cycle control. Overexpression of this phosphatase is reported to cause cell-growth arrest or cell death. Alternative splicing results in multiple transcript variants encoding different isoforms. Additional transcript variants have been described, but currently do not represent full-length sequences.

Caution: For testing in cell culture applications, please filter before use. Note that you may experience some loss of protein during the filtration process.