Order Entry
Czech Republic
ContactUsLinkComponent
Human Recombinant FOXA1
Human Recombinant FOXA1
Supplier:  OriGene
Certificates
Human Recombinant FOXA1
Supplier:  OriGene

Specifications

  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Tag sequence:
    C-Myc/DDK
  • Storage conditions:
    Store at −80 °C
  • Protein synonyms:
    HNF3A|forkhead box A1|TCF3A
  • UniProtKB:
    P55317
  • Protein/peptide name:
    FOXA1
  • Purity:
    >80% as determined by SDS-PAGE and Coomassie blue staining
  • Molecular weight:
    49 kDa
  • Sequence:
    MLGTVKMEGHETSDWNSYYADTQEAYSSVPVSNMNSGLGSMNSMNTYMTMNTMTTSGNMTPASFNMSYAN
    PGLGAGLSPGAVAGMPGGSAGAMNSMTAAGVTAMGTALSPSGMGAMGAQQAASMNGLGPYAAAMNPCMSP
    MAYAPSNLGRSRAGGGGDAKTFKRSYPHAKPPYSYISLITMAIQQAPSKMLTLSEIYQWIMDLFPYYRQN
    QQRWQNSIRHSLSFNDCFVKVARSPDKPGKGSYWTLHPDSGNMFENGCYLRRQKRFKCEKQPGAGGGGGS
    GSGGSGAKGGPESRKDPSGASNPSADSPLHRGVHGKTGQLEGAPAPGPAASPQTLDHSGATATGGASELK
    TPASSTAPPISSGPGALASVPASHPAHGLAPHESQLHLKGDPHYSFNHPFSINNLMSSSEQQHKLDFKAY
    EQALQYSPYGSTLPASLPLGSASVTTRSPIEPSALEPAYYQGVYSRPVLNTS

    TRTRPLEQKLISEEDLAANDILDYKDDDDKV
  • Concentration:
    >0,05 µg/µl as determined by microplate BCA method

Specifications

Frequently Bought Together