Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ICG hydrazide
ICG hydrazide fluorescent probe
ICG hydrazide fluorescent probe
ICG hydrazide fluorescent probe
Catalog # AATB987
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
ICG hydrazide fluorescent probe
Catalog # AATB987
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Environmentally Preferable:
Specifications
About this item
Product Details
:
:
973.04 g/mol
:
GHS07
ADR:
5 II
Product Details