Přeskočit na Záhlaví
Přeskočit na Hlavní obsah
Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cresyl violet perchlorate
Cresyl violet perchlorate ≥90%
Cresyl violet perchlorate ≥90%
AATB90
:
AAT Bioquest
:
Cresyl violet perchlorate ≥90%
AATB90
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
:
:
361.738 g/mol
:
5,II
:
GHS07
:
41830-80-2
:
C₁₆H₁₂ClN₃O₅
:
MFCD00012671