
4-Fluoro-7-nitrobenzofurazan
Catalog # AATB821
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:183.1 g/mol
ADR:5,II
:GHS07
CAS Number:29270-56-2
Formula:C₆H₂FN₃O₃
MDL Number:MFCD00010196