Order Entry
Czech Republic
ContactUsLinkComponent
5-Dimethylaminonaphthalene-1-(N-(5-aminopentyl))Sulphonamide (Dansyl cadaverine) ≥95%
5-Dimethylaminonaphthalene-1-(N-(5-aminopentyl))Sulphonamide (Dansyl cadaverine) ≥95%
Catalog # AATB810
Supplier:  AAT Bioquest
CAS Number:  
undefined
5-Dimethylaminonaphthalene-1-(N-(5-aminopentyl))Sulphonamide (Dansyl cadaverine) ≥95%
Catalog # AATB810
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :335.46 g/mol
CAS Number:10121-91-2
Formula:C₁₇H₂₅N₃O₂S
MDL Number:MFCD00042704