Order Entry
Czech Republic
ContactUsLinkComponent
3-Carboxy-6,8-difluoro-7-hydroxycoumarin ≥90% [equivalent to Pacific Blue succinimidyl ester]
3-Carboxy-6,8-difluoro-7-hydroxycoumarin ≥90% [equivalent to Pacific Blue succinimidyl ester]
Catalog # AATB570
Supplier:  AAT Bioquest
CAS Number:  
undefined
3-Carboxy-6,8-difluoro-7-hydroxycoumarin ≥90% [equivalent to Pacific Blue succinimidyl ester]
Catalog # AATB570
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :339.2 g/mol
CAS Number:215868-33-0
Formula:C₁₄H₇F₂NO₇