
FMOC-Lys(Cy5)-OH for biomolecular labeling
Catalog # AATB5055
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
ADR:5,II
:1121.24 g/mol
:GHS07
CAS Number:312961-84-5