Order Entry
Czech Republic
ContactUsLinkComponent
Cyanine 5.5 monosuccinimidyl ester potassium salt ≥90% [same as Cy5.5® NHS ester]
Cyanine 5.5 monosuccinimidyl ester potassium salt ≥90% [same as Cy5.5® NHS ester]
Catalog # AATB283
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cyanine 5.5 monosuccinimidyl ester potassium salt ≥90% [same as Cy5.5® NHS ester]
Catalog # AATB283
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :1128.4 g/mol