Order Entry
Czech Republic
ContactUsLinkComponent
Tide Quencher™ 3WS succinimidyl ester, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 3WS succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2229
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Tide Quencher™ 3WS succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2229
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :996.24 g/mol
ADR:5,II
 :GHS07