
Tide Quencher™ 2 succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2210
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:479.55 g/mol
ADR:5,II
:GHS07