Order Entry
Czech Republic
ContactUsLinkComponent
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95%
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95%
Catalog # AATB22046
Supplier:  AAT Bioquest
CAS Number:  
undefined
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95%
Catalog # AATB22046
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :572.531 g/mol
CAS Number:53213-82-4
Formula:C₂₉H₃₇IN₂O₂
MDL Number:MFCD00011954