Order Entry
Czech Republic
ContactUsLinkComponent
Tide Quencher™ 1 maleimide, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 1 maleimide, Tide Quencher™ fluorescent probe
Catalog # AATB2196
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Tide Quencher™ 1 maleimide, Tide Quencher™ fluorescent probe
Catalog # AATB2196
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :412.46 g/mol
ADR:5,II
 :GHS07