Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Zadání objednávky
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
PDMPO Dextran
PDMPO Dextran, RatioWorks™
PDMPO Dextran, RatioWorks™
Irritant
PDMPO Dextran, RatioWorks™
Catalog # AATB21211
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
PDMPO Dextran, RatioWorks™
Catalog # AATB21211
Supplier:
AAT Bioquest
CAS Number:
Specifications
Typ balení:
Plastic bottle
Bal.:
1 mg
Sekvence:
2070
Specifications
About this item
Product Details
:
:
10000 g/mol
ADR:
5,II
:
GHS07
Product Details