
Tide Quencher™ 7WS succinimidyl ester, Tide Quencher™ fluorescent probe
Catalog # AATB2111
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:1000.27 g/mol
ADR:5,II
:GHS07