Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DABCYL C2 amine
DABCYL C2 amine FRET acceptor precursor
DABCYL C2 amine FRET acceptor precursor
Irritant
DABCYL C2 amine FRET acceptor precursor
Catalog # AATB2006
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
DABCYL C2 amine FRET acceptor precursor
Catalog # AATB2006
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
:
425.41 g/mol
ADR:
5,II
:
GHS07
Product Details