
TWO-PRO™ 3 5 mM in DMSO ≥95%
Catalog # AATB17572
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:0,2 mL
- Seq:2020
Specifications
About this item
:
ADR:5,II
:671.42 g/mol
:GHS07
CAS Number:157199-63-8
Formula:C₂₆H₃₁I₂N₃S