Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in Tris/EDTA (TE) buffer ≥95%
Catalog # AATB17032
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
5-Bromouridine-5′-triphosphate sodium salt (BrUTP ) 10 mM in Tris/EDTA (TE) buffer ≥95%
Catalog # AATB17032
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 µl
  • Seq:
    1286

Specifications

About this item

 :
 :628.98 g/mol
ADR:5,II
 :GHS07
CAS Number:161848-60-8
Formula:C₉H₁₁BrN₂Na₃O₁₅P₃