Order Entry
Czech Republic
ContactUsLinkComponent
Biotin-11-dUTP 1 mM in TRIS buffer ≥95% (pH 7.5)
Irritant
Biotin-11-dUTP 1 mM in TRIS buffer ≥95% (pH 7.5)
Catalog # AATB17016
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Biotin-11-dUTP 1 mM in TRIS buffer ≥95% (pH 7.5)
Catalog # AATB17016
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 Nmo
  • Seq:
    0311

Specifications

About this item

 :
 :862.7 g/mol
ADR:5,II
 :GHS07
CAS Number:86303-25-5
Formula:C₂₈H₄₅N₆O₁₇P₃S