0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Cyanine 5 monosuccinimidyl ester
Cyanine 5 monosuccinimidyl ester ≥90% [equivalent to Cy5® NHS ester]
Cyanine 5 monosuccinimidyl ester ≥90% [equivalent to Cy5® NHS ester]
AATB151
:
AAT Bioquest
:
Cyanine 5 monosuccinimidyl ester ≥90% [equivalent to Cy5® NHS ester]
AATB151
:
AAT Bioquest
:
Pk:
1 mg
Seq:
2070
:
:
753.8814 g/mol
:
146368-14-1
:
C₃₇H₄₃N₃O₁₀S₂
:
MFCD29472578