Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Home
BOC-Val-Pro-Arg-AMC
BOC-Val-Pro-Arg-AMC
BOC-Val-Pro-Arg-AMC
Irritant
BOC-Val-Pro-Arg-AMC
Catalog # AATB13461
Supplier:
AAT BIOQUEST
CAS Number:
Rozbalit obrázek
Irritant
BOC-Val-Pro-Arg-AMC
Catalog # AATB13461
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details