Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Z-DEVD-AFC
Z-DEVD-AFC fluorogenic substrate
Z-DEVD-AFC fluorogenic substrate
AATB13420
:
AAT Bioquest
:
Z-DEVD-AFC fluorogenic substrate
AATB13420
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
:
:
821.71 g/mol
:
5,II
:
GHS07
:
1135416-11-3