Order Entry
Czech Republic
ContactUsLinkComponent
SPINeasy® DNA Pro Kit for Feces, MP Biomedicals
SPINeasy® DNA Pro Kit for Feces, MP Biomedicals
Catalog # 116547050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® DNA Pro Kit for Feces, MP Biomedicals
Catalog # 116547050.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Enzymatic extraction
  • Sample type:
    Stool
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA Pro kit for feces, 50 preps
  • Storage temperature:
    15...25 °C

Specifications

About this item

The SPINeasy® DNA Pro Kit for Feces is able to handle a wide range of fecal samples.

  • Newly formulated buffers to achieve better yield and purity of fecal DNA
  • Easy to handle and minimise any risk of contamination
  • Suitable for various types of feces samples
  • User friendly, suitable for any scale of experimental throughput

The DNA yield, purity (260/280 and 260/230 ratio) and integrity were assessed using spectrophotometer and DNA gel. Each dot of the plot represents a single extraction. The horizontal bars indicate the median value.