Mouse IgG3 Isotype Control
Catalog # NOVUNBP1-96978
Supplier: Novus Biologicals
Specifications
- Antibody type:Control
- Antigen name:HNF-3 alpha/FoxA1
- Antigen symbol:IgG3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG3
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:3502
- Antigen synonyms:FLJ40834|FLJ40036|FLJ40587|IgG3|FLJ39988|FLJ40789|FLJ40253|DKFZp686H11213|immunoglobulin heavy constant gamma 3 (G3m marker)|MGC45809
- Storage buffer:0,02 M Potassium Phosphate, 0,5 M Sodium Chloride pH 7,2
- Storage temperature:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1,0 mg/ml
- Purification:Immunoelectrophoresis (IEP)
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Mouse IgG3 Protein. Tested Applications: ELISA, Immunoprecipitation. Tested Reactivity: Mouse.