Order Entry
Czech Republic
ContactUsLinkComponent
Histamine dihydrochloride 100 µg/ml in water, colourless, liquid
Irritant
Histamine dihydrochloride 100 µg/ml in water, colourless, liquid
Catalog # EHERA14213050WL100
Supplier:  EHRENSTORFER
CAS Number:  
Irritant
Histamine dihydrochloride 100 µg/ml in water, colourless, liquid
Catalog # EHERA14213050WL100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Accurate mass:
    183.033
  • Shipping temperature:
    20 °C
  • Odour:
    Aromatic
  • Stability:
    Stable under normal conditions

Specification Test Results

About this item

 :
 :184.07 g/mol
UN:1648
ADR:3,II
 :GHS07
CAS Number:56-92-8
Formula:C₅H₁₁Cl₂N₃
MDL Number:MFCD00012703