Order Entry
Czech Republic
ContactUsLinkComponent
2,4,6-Trichloroanisole
Irritant
2,4,6-Trichloroanisole
Catalog # CMPLSB5500.100MG
CAS Number:  
Irritant
2,4,6-Trichloroanisole
Catalog # CMPLSB5500.100MG
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • XLogP3:
    4
  • Hydrogen bond donor count:
    0
  • Rotatable bond count:
    0
  • Exact mass:
    1
  • Monoisotopic mass:
    209.940598 g/mol
  • Topological polar surface area:
    209.940598 g/mol
  • Heavy atom count:
    9.2 Ų
  • Formal charge:
    11
  • Complexity:
    0
  • Isotope atom count:
    119
  • Defined atom stereocenter count:
    0
  • Undefined atom stereocenter count:
    0
  • Defined bond stereocenter count:
    0
  • Undefined bond stereocenter count:
    0
  • Covalently-bonded unit count:
    1
  • Compound is canonicalized:
    Yes
  • Hydrogen bond acceptor count:
    1

Specification Test Results

About this item

 :
 :211.47 g/mol
Boiling Pt:241 °C
Melting Pt:61...62 °C
 :GHS07
CAS Number:87-40-1
Formula:C₇H₅Cl₃O
MDL Number:MFCD00000588