IPTG (isopropyl-β-D-thiogalactopyranoside), powder
Catalog # SIALPHG0010-5G
Supplier: Merck
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Quality level:300
Specification Test Results
About this item
:
:238.31 g/mol
CAS Number:367-93-1
Formula:C₉H₁₈O₅S
MDL Number:MFCD00063273