Order Entry
Czech Republic
ContactUsLinkComponent
 
The IgG (Monkey) ELISA kit
Catalog # ABNOKA6749.1KIT
Supplier:  Abnova
CAS Number:  
The IgG (Monkey) ELISA kit
Catalog # ABNOKA6749.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Monkey
  • Target protein:
    IgG

Specifications

About this item

The IgG (Monkey) ELISA Kit is used for determination of IgG in old world monkey serum or plasma.