IgM (Sheep) ELISA kit
Catalog # ABNOKA6766.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Sheep
- Target protein:IgM
- Assay Principle:Quantitative
Specifications
About this item
IgM (Sheep) ELISA Kit is for the quantitative measurement of sheep IgM in serum or plasma.