Order Entry
Czech Republic
ContactUsLinkComponent
Echinococcus IgG ELISA kit
Echinococcus IgG ELISA kit
Catalog # ABNOKA6730.1KIT
Supplier:  Abnova
CAS Number:  
Echinococcus IgG ELISA kit
Catalog # ABNOKA6730.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    IgG
  • Assay Principle:
    Qualitative

Specifications

About this item

Echinococcus IgG ELISA Kit is a qualitative determination of IgG class antibodies against Echinococcus in human serum or plasma.