Toxoplasma IgM ELISA kit
Catalog # ABNOKA6699.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Toxoplasma gondii
- Target protein:IgM
- Assay Principle:Quantitative
Specifications
About this item
Toxoplasma IgM ELISA Kit is an enzyme immunoassay for the quantitative and qualitative measurement of Toxoplasma IgM in human serum and plasma.