Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Enzymes
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
Catalog # ABNOP9371.500UL
Supplier:
Abnova
CAS Number:
Expand image
Synthetic Yeast Inorganic Pyrophosphatase (from E. coli)
Catalog # ABNOP9371.500UL
Supplier:
Abnova
CAS Number:
Specifications
Pk:
500 µl
Enzyme type:
Synthetic
Source:
E. coli
Species:
Yeast
Enzyme name:
Inorganic Pyrophosphatase
Specifications
About this item
Product Details
Product Details