Recombinant Rat FGF21 (from E. coli)
20 µG
Katalogové číslo ABNOP8633.20UG
Dodavatel: Abnova
Přejít na:
Product Details & Documents
Specifikace
Specifikace
- Pk:20 µG
- Cat. No.:ABNOP8633.20UG
- Protein/peptide type:Recombinant
- Protein/peptide name:FGF21
- Species:Rat
- Source:E. coli
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



