Order Entry
Czech Republic
ContactUsLinkComponent
 
Recombinant Human ANGPTL4 (from E. coli), His-tag
  ABNOP8412.10UG
 :  Abnova
 :  
Recombinant Human ANGPTL4 (from E. coli), His-tag
  ABNOP8412.10UG
 :  Abnova
 :  

 

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Environmentally Preferable:
  • Tag sequence:
    His-tag
  • Gene ID:
    Q9BY76
  • Protein/peptide name:
    ANGPTL4