0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Proteins and Peptides
Recombinant Rat IGF1 (from E. coli)
Recombinant Rat IGF1 (from E. coli)
ABNOP7405.10UG
:
Abnova
:
Recombinant Rat IGF1 (from E. coli)
ABNOP7405.10UG
:
Abnova
:
Pk:
10 µG
Protein/peptide type:
Recombinant
Source:
E. coli
Species:
Rat
Environmentally Preferable:
Gene ID:
P08025
Protein/peptide name:
IGF1
Sequence:
49a.a.-118a.a.