O tomto produktu
Tygon Microbore Tubing is special application tubing used in AutoAnalyser instrumentation, ICP peristaltic pumps, and variety of flow analysis methods (segmented flow, flow injection flow batch). Major equipment and micro pump manufacturers depend on Saint-Gobain’s extensive knowledge of this specialised tubing, while scientists depend on the 2-stop and 3-stop tubing.
- Available in 2-stop and 3-stop configurations and nominal tubing
- Firm stop adhesion reduces risk of tubing slip within pumps; prevents variable flow rates to improve productivity, accuracy and safety
- Tubing available in a variety of inner diameter (ID) sizes to ensure fit
- Precision engineered with tight tolerances for optimal fluid control
- Custom assemblies are available
Tygon Microbore Tubing from Saint-Gobain is available in a range of application specific formulations with broad chemical resistance for laboratory, environmental, biopharmaceutical, clinical and industrial automated fluid sample analysis. Tygon Microbore tubing from Saint-Gobain allows optimal flow rate and throughput with accurate, reproducible results. Life science applications tubing for cell culture, reagent dispensing and microbiological testing and dilutions.
Hardness: 64 (Shore A)
Specifikace
- Catalog No:
- VERN070539-X19
- VERN070539-X15
- Min. temperature:
- −60 °C
- −60 °C
- Max. temperature:
- 135 °C
- 135 °C