Anti-KRT7 Mouse Recombinant Antibody [clone: OV-TL 12/30]
ABNORAB03399.200UG
: Abnova
- Antigen symbol:KRT7
- Clonality:Recombinant
- Clone:OV-TL 12/30
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human KRT7.
- Pk:200 µG